Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278463.1 | Intron | NP_001265392.1 | |||
NM_001278464.1 | Intron | NP_001265393.1 | |||
NM_001278465.1 | Intron | NP_001265394.1 | |||
NM_001278466.1 | Intron | NP_001265395.1 | |||
NM_005690.4 | Intron | NP_005681.2 | |||
NM_012062.4 | Intron | NP_036192.2 | |||
NM_012063.3 | Intron | NP_036193.2 | |||
XM_005253282.4 | Intron | XP_005253339.1 | |||
XM_011520543.2 | Intron | XP_011518845.1 | |||
XM_011520544.2 | Intron | XP_011518846.1 | |||
XM_017018663.1 | Intron | XP_016874152.1 | |||
XM_017018664.1 | Intron | XP_016874153.1 | |||
XM_017018665.1 | Intron | XP_016874154.1 |