Product Details

SNP ID
rs117535081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11657652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGCCTGATGGCCTCCCTGGGG[G/T]TATTTTCCCTTTGTATGTGGGAGTC
Phenotype
MIM: 600618
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ETV6 PubMed Links

Gene Details

Gene
ETV6
Gene Name
ETS variant 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001987.4 Intron NP_001978.1
XM_011520607.2 Intron XP_011518909.1
XM_011520608.2 Intron XP_011518910.1
XM_011520609.2 Intron XP_011518911.1
XM_011520611.2 Intron XP_011518913.1
XM_011520612.2 Intron XP_011518914.1
XM_017018990.1 Intron XP_016874479.1
XM_017018991.1 Intron XP_016874480.1

View Full Product Details