Product Details

SNP ID
rs116970496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12943471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGAATAATGACAGTCATAACA[A/G]TAAGTCTATCTCATAGGATTGCTTT
Phenotype
MIM: 607437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM234B PubMed Links

Gene Details

Gene
FAM234B
Gene Name
family with sequence similarity 234 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020853.1 Intron NP_065904.1
XM_017019706.1 Intron XP_016875195.1
Gene
GPRC5D
Gene Name
G protein-coupled receptor class C group 5 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018654.1 Intron NP_061124.1
XM_005253421.3 Intron XP_005253478.1
XM_017019582.1 Intron XP_016875071.1
XM_017019583.1 Intron XP_016875072.1

View Full Product Details