Product Details

SNP ID
rs115904911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123269306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTAGGTTTTGCAGGGCTGAGAGG[G/T]AGGAGGAAGGAGAAATCGTCCCATT
Phenotype
MIM: 602198
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK2AP1 PubMed Links

Gene Details

Gene
CDK2AP1
Gene Name
cyclin dependent kinase 2 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270433.1 Intron NP_001257362.1
NM_001270434.1 Intron NP_001257363.1
NM_004642.3 Intron NP_004633.1

View Full Product Details