Product Details

SNP ID
rs113195520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:131956631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTGTATGCAGTCTACTCCTAC[A/G]GGGCGTTTTTTGGGTTCTGAACCCA
Phenotype
MIM: 606265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EP400 PubMed Links

Gene Details

Gene
EP400
Gene Name
E1A binding protein p400
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015409.4 Intron NP_056224.3

View Full Product Details