Product Details

SNP ID
rs117875524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1825682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGTGGGTGTGTGTTTGCATCA[C/T]GTGCTGTGTACAGAGCTCCAAGCAC
Phenotype
MIM: 608171
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA2D4 PubMed Links

Gene Details

Gene
CACNA2D4
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172364.4 Intron NP_758952.4
XM_011521041.2 Intron XP_011519343.1
Gene
LRTM2
Gene Name
leucine rich repeats and transmembrane domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039029.2 Intron NP_001034118.1
NM_001163925.1 Intron NP_001157397.1
NM_001163926.1 Intron NP_001157398.1
XM_011521015.1 Intron XP_011519317.1
XM_017019848.1 Intron XP_016875337.1

View Full Product Details