Product Details

SNP ID
rs114779646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4321687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCAAAGAGTTTGCAGAATTGCT[C/T]GTCCATGGCTGATGTTTTTGATGCC
Phenotype
MIM: 610775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIGAR PubMed Links

Gene Details

Gene
TIGAR
Gene Name
TP53 induced glycolysis regulatory phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020375.2 Intron NP_065108.1

View Full Product Details