Product Details

SNP ID
rs112235025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6981003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTGGACCAAGAGGGGCAATTA[C/G]CTGTTTGGTATCTTTCCTGGTATGT
Phenotype
MIM: 611531 MIM: 611950
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMG1 PubMed Links

Gene Details

Gene
EMG1
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320049.1 Intron NP_001306978.1
NM_006331.7 Intron NP_006322.4
Gene
LPCAT3
Gene Name
lysophosphatidylcholine acyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005768.5 Intron NP_005759.4

View Full Product Details