Product Details
- SNP ID
-
rs112677599
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:913903 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACACCTCTCTGCTACCTGTTGTGC[A/G]TTGGTCAGCGCTATAAGTTTGGAGG
- Phenotype
-
MIM: 600392
MIM: 605232
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RAD52
PubMed Links
Gene Details
- Gene
- RAD52
- Gene Name
- RAD52 homolog, DNA repair protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001297419.1 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
NP_001284348.1 |
NM_001297420.1 |
1582 |
Intron |
|
|
NP_001284349.1 |
NM_001297421.1 |
1582 |
Missense Mutation |
CGC,TGC |
R319C |
NP_001284350.1 |
NM_001297422.1 |
1582 |
Intron |
|
|
NP_001284351.1 |
NM_134424.3 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
NP_602296.2 |
XM_005253720.4 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
XP_005253777.1 |
XM_005253721.2 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
XP_005253778.1 |
XM_011520990.2 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
XP_011519292.1 |
XM_011520991.2 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
XP_011519293.1 |
XM_011520993.2 |
1582 |
Intron |
|
|
XP_011519295.1 |
XM_011520995.2 |
1582 |
Missense Mutation |
CGC,TGC |
R298C |
XP_011519297.1 |
XM_017019769.1 |
1582 |
Missense Mutation |
CGC,TGC |
R396C |
XP_016875258.1 |
XM_017019770.1 |
1582 |
Missense Mutation |
CGC,TGC |
R362C |
XP_016875259.1 |
XM_017019771.1 |
1582 |
Intron |
|
|
XP_016875260.1 |
- Gene
- WNK1
- Gene Name
- WNK lysine deficient protein kinase 1
There are no transcripts associated with this gene.
View Full Product Details