Product Details

SNP ID
rs112613649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120450088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTTCTTTATTAATATCTCTAG[A/G]AAAGGAGGGACTAGAAGCACAGAGT
Phenotype
MIM: 602072 MIM: 614943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX6A1 PubMed Links

Gene Details

Gene
COX6A1
Gene Name
cytochrome c oxidase subunit 6A1
There are no transcripts associated with this gene.

Gene
GATC
Gene Name
glutamyl-tRNA amidotransferase subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176818.2 Intron NP_789788.1
Gene
TRIAP1
Gene Name
TP53 regulated inhibitor of apoptosis 1
There are no transcripts associated with this gene.

View Full Product Details