Product Details

SNP ID
rs114352080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121217108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCCATTGAATCCTAATGGCAA[C/T]TTAAGAGGCCTGTTTTATTTTATAG
Phenotype
MIM: 600846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RX4 PubMed Links

Gene Details

Gene
P2RX4
Gene Name
purinergic receptor P2X 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256796.1 Intron NP_001243725.1
NM_001261397.1 Intron NP_001248326.1
NM_001261398.1 Intron NP_001248327.1
NM_002560.2 Intron NP_002551.2
XM_011538416.2 Intron XP_011536718.1

View Full Product Details