Product Details

SNP ID
rs113279920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121810045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACTGGGGGCAGCAGCGGTCCCC[C/T]GTTCAAGGCTCAACCACAGGATTCA
Phenotype
MIM: 611055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01089 PubMed Links

Gene Details

Gene
LINC01089
Gene Name
long intergenic non-protein coding RNA 1089
There are no transcripts associated with this gene.

Gene
SETD1B
Gene Name
SET domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015048.1 1265 Missense Mutation CCG,CTG P367L NP_055863.1
XM_005253858.4 1265 Missense Mutation CCG,CTG P367L XP_005253915.1
XM_006719296.3 1265 Missense Mutation CCG,CTG P367L XP_006719359.1

View Full Product Details