Product Details

SNP ID
rs114642524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111686141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGGTGCCACACTTAGGCTGAG[C/T]TGCAGGTTTTCGCACAGTCGCGAGT
Phenotype
MIM: 611181 MIM: 604986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAD10 PubMed Links

Gene Details

Gene
ACAD10
Gene Name
acyl-CoA dehydrogenase family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136538.1 89 UTR 5 NP_001130010.1
NM_025247.5 89 UTR 5 NP_079523.3
Gene
BRAP
Gene Name
BRCA1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006768.4 89 Intron NP_006759.3
XM_005253944.4 89 Intron XP_005254001.1
XM_011538789.2 89 Intron XP_011537091.1
XM_017019992.1 89 Intron XP_016875481.1
XM_017019993.1 89 Intron XP_016875482.1
XM_017019994.1 89 Intron XP_016875483.1
XM_017019995.1 89 Intron XP_016875484.1

View Full Product Details