Product Details

SNP ID
rs115609511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113159554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTATGTCTCCACAGCACACCTA[C/T]GGCACGCCAGGCTCTGAGGCCCCCC
Phenotype
MIM: 611665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP73 PubMed Links

Gene Details

Gene
CFAP73
Gene Name
cilia and flagella associated protein 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144872.1 Intron NP_001138344.1
XM_011538327.2 Intron XP_011536629.1
Gene
DDX54
Gene Name
DEAD-box helicase 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111322.1 Intron NP_001104792.1
NM_024072.3 Intron NP_076977.3
Gene
MIR7106
Gene Name
microRNA 7106
There are no transcripts associated with this gene.

View Full Product Details