Product Details

SNP ID
rs114256458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116719851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGGAATGCCACGGCTGCCCGG[C/T]TGAGGAAGCGCTCCAGGAGAAGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf49 PubMed Links

Gene Details

Gene
C12orf49
Gene Name
chromosome 12 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024738.2 616 Missense Mutation AAC,AGC N149S NP_079014.1
XM_005253937.3 616 Missense Mutation AAC,AGC N119S XP_005253994.1

View Full Product Details