Product Details

SNP ID
rs118121557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:117043191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGGAGAAAATGGGAAGCCACATC[A/G]GTTCGATTATGTTTTGCATACATCA
Phenotype
MIM: 611585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TESC PubMed Links

Gene Details

Gene
TESC
Gene Name
tescalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168325.1 Intron NP_001161797.1
NM_017899.3 Intron NP_060369.3
XM_011538503.1 Intron XP_011536805.1
XM_017019532.1 Intron XP_016875021.1
XM_017019533.1 Intron XP_016875022.1

View Full Product Details