Product Details

SNP ID
rs112716013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110715615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACGAGTTGATTTGAACGTCTTC[G/T]GGTCGCTCGGCCTCCAGCCTTGGAT
Phenotype
MIM: 612800 MIM: 601566
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CARS2 PubMed Links

Gene Details

Gene
CARS2
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
There are no transcripts associated with this gene.

Gene
ING1
Gene Name
inhibitor of growth family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267728.1 604 Intron NP_001254657.1
NM_005537.5 604 Missense Mutation GGG,TGG G58W NP_005528.4
NM_198217.2 604 Intron NP_937860.1
NM_198218.2 604 Intron NP_937861.1
NM_198219.2 604 Intron NP_937862.1
Gene
LOC105376684
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details