Product Details

SNP ID
rs117057165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15643791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTGTGCAAAGACTCAACTGGAC[A/G]TTGCCACCAAGAATCAAGAGATTTT
Phenotype
MIM: 614593 MIM: 609449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0430 PubMed Links

Gene Details

Gene
KIAA0430
Gene Name
KIAA0430
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184998.1 525 Intron NP_001171927.1
NM_001184999.1 525 Intron NP_001171928.1
NM_014647.3 525 Intron NP_055462.2
XM_005255704.1 525 Intron XP_005255761.1
XM_005255706.1 525 Intron XP_005255763.1
XM_005255707.1 525 Intron XP_005255764.1
XM_017023900.1 525 Intron XP_016879389.1
XM_017023901.1 525 Intron XP_016879390.1
XM_017023902.1 525 Intron XP_016879391.1
XM_017023903.1 525 Intron XP_016879392.1
XM_017023904.1 525 Intron XP_016879393.1
XM_017023905.1 525 Intron XP_016879394.1
XM_017023906.1 525 Intron XP_016879395.1
Gene
MIR484
Gene Name
microRNA 484
There are no transcripts associated with this gene.

Gene
NDE1
Gene Name
nudE neurodevelopment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143979.1 525 UTR 5 NP_001137451.1
NM_017668.2 525 Intron NP_060138.1
XM_005255396.4 525 Intron XP_005255453.1
XM_006720897.3 525 UTR 5 XP_006720960.1
XM_006720900.3 525 Intron XP_006720963.1
XM_011522553.2 525 UTR 5 XP_011520855.1
XM_017023349.1 525 Intron XP_016878838.1
XM_017023350.1 525 Intron XP_016878839.1
XM_017023351.1 525 Intron XP_016878840.1
XM_017023352.1 525 Intron XP_016878841.1
XM_017023353.1 525 Intron XP_016878842.1
XM_017023354.1 525 Intron XP_016878843.1
XM_017023355.1 525 Intron XP_016878844.1
XM_017023356.1 525 Intron XP_016878845.1
XM_017023357.1 525 Intron XP_016878846.1

View Full Product Details