Product Details

SNP ID
rs116253586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:17104235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAGAGCCTGGCTGTTTCACAC[C/G]CTACCTCCTCCTCCTCCTAGACTGT
Phenotype
MIM: 608124
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XYLT1 PubMed Links

Gene Details

Gene
XYLT1
Gene Name
xylosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022166.3 7425 UTR 3 NP_071449.1
XM_017023539.1 7425 Intron XP_016879028.1
XM_017023540.1 7425 Intron XP_016879029.1

View Full Product Details