Product Details

SNP ID
rs115041729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:189494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCAACCAGACTTGCCCACCAC[A/G]TATGGAGTGTTAGATAGCCATTAAA
Phenotype
MIM: 142240 MIM: 607782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HBQ1 PubMed Links

Gene Details

Gene
HBQ1
Gene Name
hemoglobin subunit theta 1
There are no transcripts associated with this gene.

Gene
LUC7L
Gene Name
LUC7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320226.1 1591 Intron NP_001307155.1
NM_018032.4 1591 UTR 3 NP_060502.1
NM_201412.2 1591 Intron NP_958815.1
XM_005255427.3 1591 Intron XP_005255484.1
XM_005255429.3 1591 Intron XP_005255486.1
XM_011522561.2 1591 Intron XP_011520863.1
XM_017023437.1 1591 Intron XP_016878926.1
XM_017023438.1 1591 Intron XP_016878927.1
XM_017023439.1 1591 Intron XP_016878928.1
XM_017023440.1 1591 Intron XP_016878929.1

View Full Product Details