Product Details

SNP ID
rs116496443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19115148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGGCCACTGGCTTCGGGACTG[C/T]AAACCCTTTGCTGATGCCTTCTGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITPRIPL2 PubMed Links

Gene Details

Gene
ITPRIPL2
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034841.3 1217 Silent Mutation TGC,TGT C229C NP_001030013.1

View Full Product Details