Product Details

SNP ID
rs117906116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20766072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAATAAATAAATGAATCATGCCA[A/G]CCTACCAAGAATGAAGCTCAAGACC
Phenotype
MIM: 145505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSM3 PubMed Links

Gene Details

Gene
ACSM3
Gene Name
acyl-CoA synthetase medium-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005622.3 Intron NP_005613.2
NM_202000.2 Intron NP_973729.1
XM_011545911.1 Intron XP_011544213.1
XM_017023523.1 Intron XP_016879012.1
XM_017023524.1 Intron XP_016879013.1

View Full Product Details