Product Details

SNP ID
rs111820849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21197490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTAGAAGCCCATTTAGTGATTT[A/G]ACACAGTCCTTTTCTCGTACAGGTA
Phenotype
MIM: 182888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZP2 PubMed Links

Gene Details

Gene
ZP2
Gene Name
zona pellucida glycoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290104.1 2684 Missense Mutation TCA,TTA S734L NP_001277033.1
NM_003460.2 2684 Missense Mutation TCA,TTA S743L NP_003451.1
XM_011545942.2 2684 Missense Mutation TCA,TTA S754L XP_011544244.1

View Full Product Details