Product Details

SNP ID
rs117420101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21260815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACAGGGCTTAGGCTATACTTTC[C/T]ATCTGACTCTTTCAGGTTGGGTCTC
Phenotype
MIM: 609901 MIM: 123740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS4B PubMed Links

Gene Details

Gene
ANKS4B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
There are no transcripts associated with this gene.

Gene
CRYM
Gene Name
crystallin mu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001888.4 Intron NP_001879.1

View Full Product Details