Product Details

SNP ID
rs117991403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22109708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCTTATAAAGAACAGTTCACA[A/C]TTAATTAGCACCTGTGGTTAGAGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VWA3A PubMed Links

Gene Details

Gene
VWA3A
Gene Name
von Willebrand factor A domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173615.4 Intron NP_775886.3
XM_011545742.2 Intron XP_011544044.1
XM_011545744.2 Intron XP_011544046.1
XM_011545745.1 Intron XP_011544047.1
XM_011545746.2 Intron XP_011544048.1
XM_017022947.1 Intron XP_016878436.1
XM_017022948.1 Intron XP_016878437.1
XM_017022949.1 Intron XP_016878438.1

View Full Product Details