Product Details

SNP ID
rs117998954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23647934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAACCCTATACCCAACTGAAGAA[C/T]TGGAACATAATCTGTATACAATAAG
Phenotype
MIM: 612962 MIM: 610355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCTN5 PubMed Links

Gene Details

Gene
DCTN5
Gene Name
dynactin subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199011.1 Intron NP_001185940.1
NM_001199743.1 Intron NP_001186672.1
NM_032486.3 Intron NP_115875.1
Gene
PALB2
Gene Name
partner and localizer of BRCA2
There are no transcripts associated with this gene.

View Full Product Details