Product Details

SNP ID
rs114176225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:281850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGGAAGAGCCTCTTGGAGATC[C/T]GGCAGCTGGACCCGGACGACAGGAG
Phenotype
MIM: 602844 MIM: 603816 MIM: 608012 MIM: 603895
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGDIG PubMed Links

Gene Details

Gene
ARHGDIG
Gene Name
Rho GDP dissociation inhibitor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001176.3 253 Missense Mutation CGG,TGG R60W NP_001167.2
Gene
AXIN1
Gene Name
axin 1
There are no transcripts associated with this gene.

Gene
PDIA2
Gene Name
protein disulfide isomerase family A member 2
There are no transcripts associated with this gene.

Gene
RGS11
Gene Name
regulator of G-protein signaling 11
There are no transcripts associated with this gene.

View Full Product Details