Product Details
- SNP ID
-
rs111720341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28511269 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGGGCTGAGGCCTGGGTTAGTG[A/C]CAGCAGGAAATGGAAGGAAGGTAGT
- Phenotype
-
MIM: 608273
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL27
PubMed Links
Gene Details
- Gene
- IL27
- Gene Name
- interleukin 27
View Full Product Details