Product Details

SNP ID
rs115303964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30023689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTCGAGAGATAAAGGCAGGGGT[A/C]AGCTTGGCTGTTGTCACAGAGTTTG
Phenotype
MIM: 604567 MIM: 615175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C16orf92 PubMed Links

Gene Details

Gene
C16orf92
Gene Name
chromosome 16 open reading frame 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109659.1 114 Intron NP_001103129.1
NM_001109660.1 114 Silent Mutation GTA,GTC V31V NP_001103130.1
Gene
DOC2A
Gene Name
double C2 domain alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282062.1 114 Intron NP_001268991.1
NM_001282063.1 114 Intron NP_001268992.1
NM_001282068.1 114 Intron NP_001268997.1
NM_003586.2 114 Intron NP_003577.2
XM_011545975.1 114 Intron XP_011544277.1
XM_017023776.1 114 Intron XP_016879265.1
XM_017023777.1 114 Intron XP_016879266.1
XM_017023778.1 114 Intron XP_016879267.1
Gene
FAM57B
Gene Name
family with sequence similarity 57 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318504.1 114 Intron NP_001305433.1
NM_031478.5 114 Intron NP_113666.2
XM_005255613.3 114 Intron XP_005255670.1
XM_005255614.3 114 Intron XP_005255671.1
XM_005255615.2 114 Intron XP_005255672.1
XM_017023749.1 114 Intron XP_016879238.1
XM_017023750.1 114 Intron XP_016879239.1
XM_017023751.1 114 Intron XP_016879240.1
XM_017023752.1 114 Intron XP_016879241.1
XM_017023753.1 114 Intron XP_016879242.1
XM_017023754.1 114 Intron XP_016879243.1

View Full Product Details