Product Details

SNP ID
rs116838618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30054503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGGTAAAGGAGTGGCAAGGGA[A/G]TCAGAGACAAGAGATCTCTTTGGAG
Phenotype
MIM: 103850 MIM: 615175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDOA PubMed Links

Gene Details

Gene
ALDOA
Gene Name
aldolase, fructose-bisphosphate A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000034.3 Intron NP_000025.1
NM_001127617.2 Intron NP_001121089.1
NM_001243177.1 Intron NP_001230106.1
NM_184041.2 Intron NP_908930.1
NM_184043.2 Intron NP_908932.1
XM_011545768.2 Intron XP_011544070.1
Gene
FAM57B
Gene Name
family with sequence similarity 57 member B
There are no transcripts associated with this gene.

View Full Product Details