Product Details

SNP ID
rs116570341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31357923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGTGTGCGTGTGCACATGCAG[A/G]TTGAGACGCAGGGCCTGACTTCCCT
Phenotype
MIM: 151510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGAX PubMed Links

Gene Details

Gene
ITGAX
Gene Name
integrin subunit alpha X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000887.4 Intron NP_000878.2
NM_001286375.1 Intron NP_001273304.1
XM_011545852.1 Intron XP_011544154.1
XM_011545854.1 Intron XP_011544156.1

View Full Product Details