Product Details

SNP ID
rs113118905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3305477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGCGCCACCCTCGGAAAGCGT[C/T]ACTTCCACTTCCGGCCGGCGCTCTG
Phenotype
MIM: 612969 MIM: 604191 MIM: 601473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIGD7 PubMed Links

Gene Details

Gene
TIGD7
Gene Name
tigger transposable element derived 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033208.3 72 Intron NP_149985.2
Gene
ZNF263
Gene Name
zinc finger protein 263
There are no transcripts associated with this gene.

Gene
ZNF75A
Gene Name
zinc finger protein 75a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302109.1 72 UTR 5 NP_001289038.1
NM_001324041.1 72 Intron NP_001310970.1
NM_001324085.1 72 UTR 5 NP_001311014.1
NM_153028.3 72 UTR 5 NP_694573.1
XM_011522646.2 72 UTR 5 XP_011520948.1
XM_011522647.2 72 Intron XP_011520949.1
XM_017023642.1 72 Intron XP_016879131.1
XM_017023643.1 72 UTR 5 XP_016879132.1
XM_017023644.1 72 UTR 5 XP_016879133.1
XM_017023645.1 72 UTR 5 XP_016879134.1

View Full Product Details