Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015041.2 | 3532 | UTR 3 | NP_055856.1 | ||
NM_024793.2 | 3532 | UTR 3 | NP_079069.1 | ||
XM_006720867.3 | 3532 | Intron | XP_006720930.1 | ||
XM_017023068.1 | 3532 | Intron | XP_016878557.1 | ||
XM_017023069.1 | 3532 | Intron | XP_016878558.1 | ||
XM_017023070.1 | 3532 | Intron | XP_016878559.1 | ||
XM_017023071.1 | 3532 | Intron | XP_016878560.1 | ||
XM_017023072.1 | 3532 | Intron | XP_016878561.1 | ||
XM_017023073.1 | 3532 | Intron | XP_016878562.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_178844.3 | 3532 | Intron | NP_849172.2 | ||
XM_017023027.1 | 3532 | UTR 3 | XP_016878516.1 | ||
XM_017023028.1 | 3532 | UTR 3 | XP_016878517.1 | ||
XM_017023029.1 | 3532 | UTR 3 | XP_016878518.1 | ||
XM_017023030.1 | 3532 | UTR 3 | XP_016878519.1 | ||
XM_017023031.1 | 3532 | UTR 3 | XP_016878520.1 | ||
XM_017023032.1 | 3532 | UTR 3 | XP_016878521.1 | ||
XM_017023033.1 | 3532 | UTR 3 | XP_016878522.1 | ||
XM_017023034.1 | 3532 | UTR 3 | XP_016878523.1 | ||
XM_017023035.1 | 3532 | UTR 3 | XP_016878524.1 | ||
XM_017023036.1 | 3532 | UTR 3 | XP_016878525.1 | ||
XM_017023037.1 | 3532 | UTR 3 | XP_016878526.1 | ||
XM_017023038.1 | 3532 | UTR 3 | XP_016878527.1 | ||
XM_017023039.1 | 3532 | Intron | XP_016878528.1 |