Product Details

SNP ID
rs117431409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4512416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAATTTTTGCCAGAAGAGTCAG[C/T]GGCTCAGGTAGGGCAGGGTGAAGAG
Phenotype
MIM: 610503 MIM: 141251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDIP1 PubMed Links

Gene Details

Gene
CDIP1
Gene Name
cell death-inducing p53 target 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199054.1 764 UTR 3 NP_001185983.1
NM_001199055.1 764 UTR 3 NP_001185984.1
NM_001199056.1 764 UTR 3 NP_001185985.1
NM_013399.2 764 UTR 3 NP_037531.2
Gene
HMOX2
Gene Name
heme oxygenase 2
There are no transcripts associated with this gene.

View Full Product Details