Product Details

SNP ID
rs111511139
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56434824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGTAATATGTGCAGGAATATA[A/T]GGATACTGAAATTACAAATGAATAC
Phenotype
MIM: 603243 MIM: 604978
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
AMFR PubMed Links

Gene Details

Gene
AMFR
Gene Name
autocrine motility factor receptor
There are no transcripts associated with this gene.

Gene
NUDT21
Gene Name
nudix hydrolase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007006.2 624 Silent Mutation CCA,CCT P159P NP_008937.1

View Full Product Details