Product Details

SNP ID
rs117336532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56437630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCTTATCCAATGGGGTATAGGA[A/G]TCCCTACATGGTTCCTGGTCCTTAA
Phenotype
MIM: 604978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT21 PubMed Links

Gene Details

Gene
NUDT21
Gene Name
nudix hydrolase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007006.2 Intron NP_008937.1

View Full Product Details