Product Details

SNP ID
rs117240233
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56456616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATACAGGCAATCGCCACATGATA[C/T]TTTCTGTCAGCAACAGATACGATAG
Phenotype
MIM: 604978 MIM: 615857
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NUDT21 PubMed Links

Gene Details

Gene
NUDT21
Gene Name
nudix hydrolase 21
There are no transcripts associated with this gene.

Gene
OGFOD1
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324357.1 Intron NP_001311286.1
NM_001324358.1 Intron NP_001311287.1
NM_001324359.1 Intron NP_001311288.1
NM_001324360.1 Intron NP_001311289.1
NM_001324361.1 Intron NP_001311290.1
NM_001324362.1 Intron NP_001311291.1
NM_001324363.1 Intron NP_001311292.1
NM_018233.3 Intron NP_060703.3

View Full Product Details