Product Details

SNP ID
rs112033454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57883920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAAGGTAGGGCTCTCAAATGA[C/T]AGTGAGAGCTCAGGGACTCGAACAC
Phenotype
MIM: 600724
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNGB1 PubMed Links

Gene Details

Gene
CNGB1
Gene Name
cyclic nucleotide gated channel beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135639.1 4065 Intron NP_001129111.1
NM_001286130.1 4065 UTR 3 NP_001273059.1
NM_001297.4 4065 UTR 3 NP_001288.3
XM_011522870.2 4065 UTR 3 XP_011521172.1

View Full Product Details