Product Details

SNP ID
rs112723051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58116114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAAGGAAGTTTAATTCCCAGC[A/G]TCTTCTTGGGGTCTGCAGGGCATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP20 PubMed Links

Gene Details

Gene
CFAP20
Gene Name
cilia and flagella associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013242.2 480 Missense Mutation ACG,ATG T68M NP_037374.1

View Full Product Details