Product Details

SNP ID
rs121434387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69331067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGTTCACATGCCCTAGGTTACC[G/C]TACTTGGAGAGCTGAGTGGGAGGAA
Phenotype
MIM: 606979 MIM: 609982
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
COG8 PubMed Links

Gene Details

Gene
COG8
Gene Name
component of oligomeric golgi complex 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032382.4 1682 Silent Mutation TAC,TAG Y537* NP_115758.3
Gene
NIP7
Gene Name
NIP7, nucleolar pre-rRNA processing protein
There are no transcripts associated with this gene.

Gene
PDF
Gene Name
peptide deformylase (mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022341.1 1682 Intron NP_071736.1
Gene
VPS4A
Gene Name
vacuolar protein sorting 4 homolog A
There are no transcripts associated with this gene.

View Full Product Details