Product Details

SNP ID
rs117551417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46707683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCTCAGTAATTTCTGGACCCA[C/T]TGGAGCAGCAGAGTTGAAGATTAGG
Phenotype
MIM: 612147 MIM: 607213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYLK3 PubMed Links

Gene Details

Gene
MYLK3
Gene Name
myosin light chain kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308301.1 1778 UTR 3 NP_001295230.1
NM_182493.2 1778 UTR 3 NP_872299.2
XM_006721334.3 1778 UTR 3 XP_006721397.1
Gene
ORC6
Gene Name
origin recognition complex subunit 6
There are no transcripts associated with this gene.

View Full Product Details