Product Details

SNP ID
rs111964475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87394689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTCTCCAGTAGGTAGCCATCA[C/T]TTCCAGTATATTAAGTGGACCAAGA
Phenotype
MIM: 609102 MIM: 609604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO31 PubMed Links

Gene Details

Gene
FBXO31
Gene Name
F-box protein 31
There are no transcripts associated with this gene.

Gene
MAP1LC3B
Gene Name
microtubule associated protein 1 light chain 3 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022818.4 Intron NP_073729.1

View Full Product Details