Product Details

SNP ID
rs114061800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:51137704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTCTATTCTCATCATGTGACA[C/G]ACAGTGAAGGCAACAGGAACCATCC
Phenotype
MIM: 602218
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371253 PubMed Links

Gene Details

Gene
LOC105371253
Gene Name
uncharacterized LOC105371253
There are no transcripts associated with this gene.

Gene
SALL1
Gene Name
spalt like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127892.1 Intron NP_001121364.1
NM_002968.2 Intron NP_002959.2
XM_006721241.3 Intron XP_006721304.1
XM_011523254.2 Intron XP_011521556.1

View Full Product Details