Product Details
- SNP ID
-
rs115368866
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:53704196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTTAGTGGCAGCATGAAGCGCAC[C/G]CCGACTGCCGAGGAACGAGAGCGCG
- Phenotype
-
MIM: 610966
MIM: 610937
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FTO
PubMed Links
Gene Details
- Gene
- FTO
- Gene Name
- fat mass and obesity associated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080432.2 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
NP_001073901.1 |
XM_011523313.2 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_011521615.1 |
XM_011523314.2 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_011521616.1 |
XM_011523315.2 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_011521617.1 |
XM_011523316.2 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_011521618.1 |
XM_017023654.1 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_016879143.1 |
XM_017023655.1 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_016879144.1 |
XM_017023656.1 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_016879145.1 |
XM_017023657.1 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_016879146.1 |
XM_017023658.1 |
198 |
Silent Mutation |
ACC,ACG |
T4T |
XP_016879147.1 |
- Gene
- RPGRIP1L
- Gene Name
- RPGRIP1 like
View Full Product Details