Product Details

SNP ID
rs116512596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:54925137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGGATGAATTTCAGCCAACAT[C/T]TGGAGGAACCCCTTAGTTCAGAAGT
Phenotype
MIM: 615624 MIM: 606195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRNDE PubMed Links

Gene Details

Gene
CRNDE
Gene Name
colorectal neoplasia differentially expressed (non-protein coding)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308963.1 1495 UTR 5 NP_001295892.1
XM_017023562.1 1495 UTR 5 XP_016879051.1
XM_017023563.1 1495 UTR 5 XP_016879052.1
Gene
IRX5
Gene Name
iroquois homeobox 5
There are no transcripts associated with this gene.

View Full Product Details