Product Details

SNP ID
rs117557103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88707675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTCCCCCAGGCGCTTGTCATGT[C/T]TTTCTTGGCTTGTAGCCATTGTATT
Phenotype
MIM: 611184
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTU2 PubMed Links

Gene Details

Gene
CTU2
Gene Name
cytosolic thiouridylase subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012759.2 Intron NP_001012777.1
NM_001012762.2 Intron NP_001012780.1
NM_001318507.1 Intron NP_001305436.1
NM_001318513.1 Intron NP_001305442.1
XM_017023210.1 Intron XP_016878699.1
Gene
MIR4722
Gene Name
microRNA 4722
There are no transcripts associated with this gene.

Gene
PIEZO1
Gene Name
piezo type mechanosensitive ion channel component 1
There are no transcripts associated with this gene.

Gene
RNF166
Gene Name
ring finger protein 166
There are no transcripts associated with this gene.

View Full Product Details