Product Details

SNP ID
rs114847933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82019062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCTGCCCCTTCTGCACAGGCTG[A/G]AGACTCCCAGAGATCTTGTTTGAAT
Phenotype
MIM: 606236 MIM: 615128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251888.1 383 Intron NP_001238817.1
NM_024083.3 383 Intron NP_076988.1
XM_011523601.2 383 Intron XP_011521903.1
XM_011523602.2 383 Intron XP_011521904.1
XM_017025036.1 383 Intron XP_016880525.1
XM_017025037.1 383 Intron XP_016880526.1
XM_017025038.1 383 Intron XP_016880527.1
XM_017025039.1 383 Intron XP_016880528.1
XM_017025040.1 383 Intron XP_016880529.1
XM_017025041.1 383 Intron XP_016880530.1
XM_017025042.1 383 Intron XP_016880531.1
XM_017025043.1 383 Intron XP_016880532.1
XM_017025044.1 383 Intron XP_016880533.1
Gene
LRRC45
Gene Name
leucine rich repeat containing 45
There are no transcripts associated with this gene.

Gene
STRA13
Gene Name
stimulated by retinoic acid 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271006.1 383 UTR 3 NP_001257935.1
NM_001271007.1 383 UTR 3 NP_001257936.1
NM_144998.3 383 UTR 3 NP_659435.2
XM_005256339.2 383 UTR 3 XP_005256396.1
XM_017024326.1 383 UTR 3 XP_016879815.1
XM_017024327.1 383 UTR 3 XP_016879816.1
XM_017024328.1 383 UTR 3 XP_016879817.1
XM_017024329.1 383 UTR 3 XP_016879818.1

View Full Product Details