Product Details

SNP ID
rs116114329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82362299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTAGACTTTAAAGACTTGCTGG[A/C]GTCAGAGGACTGGGAAGAAGACAAC
Phenotype
MIM: 615647
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TEX19 PubMed Links

Gene Details

Gene
TEX19
Gene Name
testis expressed 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207459.3 546 Missense Mutation GAG,GCG E50A NP_997342.1
XM_017024638.1 546 Missense Mutation GAG,GCG E50A XP_016880127.1
XM_017024639.1 546 Missense Mutation GAG,GCG E50A XP_016880128.1

View Full Product Details