Product Details

SNP ID
rs113265042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82362499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTTGGAAGATGCAGGTCTGGAC[C/T]CCCACTTTGTCCCCACTGAACTATG
Phenotype
MIM: 615647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX19 PubMed Links

Gene Details

Gene
TEX19
Gene Name
testis expressed 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207459.3 746 Missense Mutation CCC,TCC P117S NP_997342.1
XM_017024638.1 746 Missense Mutation CCC,TCC P117S XP_016880127.1
XM_017024639.1 746 Missense Mutation CCC,TCC P117S XP_016880128.1

View Full Product Details