Product Details

SNP ID
rs116293085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82362538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGAACTATGGCCTCAGGAGGCT[A/G]TGCCCCTGGGCCTGGGCCTTGAGGA
Phenotype
MIM: 615647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX19 PubMed Links

Gene Details

Gene
TEX19
Gene Name
testis expressed 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207459.3 785 Missense Mutation ATG,GTG M130V NP_997342.1
XM_017024638.1 785 Missense Mutation ATG,GTG M130V XP_016880127.1
XM_017024639.1 785 Missense Mutation ATG,GTG M130V XP_016880128.1

View Full Product Details